FluoCells™ Prepared Slide #2 (BPAE cells with Mouse Anti-α-tubulin, AlexaFluor™ 488, FL Goat Anti-Mouse IgG, Texas Red™-X Phalloidin, and DAPI)
FluoCells™ Prepared Slide #2 (BPAE cells with Mouse Anti-α-tubulin, AlexaFluor™ 488, FL Goat Anti-Mouse IgG, Texas Red™-X Phalloidin, and DAPI)
Invitrogen™

FluoCells™ Prepared Slide #2 (BPAE cells with Mouse Anti-α-tubulin, AlexaFluor™ 488, FL Goat Anti-Mouse IgG, Texas Red™-X Phalloidin, and DAPI)

FluoCells prepared microscope slides contain multi-labeled cell preparations for observation by fluorescence microscopy. FluoCells Prepared Slide #2 contains bovine pulmonaryRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
F14781Promo Image1 Each
Catalog number F14781
Price (USD)
293.65
Online Exclusive
317.00
Save 23.35 (7%)
Each
In stock
Add to cart
Quantity:
1 Each
Price (USD)
293.65
Online Exclusive
317.00
Save 23.35 (7%)
Each
Add to cart
FluoCells prepared microscope slides contain multi-labeled cell preparations for observation by fluorescence microscopy. FluoCells Prepared Slide #2 contains bovine pulmonary artery endothelial cells (BPAEC). Within each cell, F-actin is labelled with Texas Red-X phalloidin, microtubules are labelled with anti–bovine α-tubulin mouse monoclonal 236-10501 in conjunction with AlexaFluor 488 goat anti–mouse IgG antibody, and the nucleus is labelled with DAPI.

See our full collection of microscope calibration reagents ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Immunofluorescence
Quantity1 Each
Shipping ConditionRoom Temperature
Product LineBODIPY FluoCells Texas Red
TypePrepared Microscope Slide
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection