Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phusion Human Specimen Direct PCR Kit

Catalog number: F150BID
Have Questions?
Thermo Scientific™

Phusion Human Specimen Direct PCR Kit

Catalog number: F150BID
Have Questions?
Catalog Number
F150BID
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F150BID200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Phusion Human Specimen Direct PCR Kit delivers unprecedented convenience for DNA amplification by allowing PCR directly from unpurified human samples. A tiny amount of sample is used directly in the PCR reaction with no prior purification steps. This allows significant savings in both time and cost.

Tested sample materials include buccal swabs, saliva, amniotic fluid, hair, fingernails, teeth, and skin biopsies. The kit is compatible with fresh, frozen, and even Formalin-Fixed, Paraffin Embedded (FFPE) human tissue samples. It includes optimized reagents and detailed protocols for many sample materials. The modified Phusion Hot Start II High-Fidelity DNA Polymerase utilized in the kit is a highly accurate proofreading DNA polymerase; PCR products obtained are perfectly suitable even for cloning purposes.

Highlights

• No need for time-consuming and expensive DNA purification steps
• Two simple protocols for various applications
• Extremely short PCR protocol times
• Modified Phusion Hot Start II High-Fidelity DNA Polymerase delivers abundant yields of specific product

The optimal annealing temperature for Phusion DNA Polymerases may differ significantly from that of Taq-based DNA polymerases.

For optimal results start by accurately calculating your Tm with our Tm calculator.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
PCR Method
Direct PCR
Polymerase
Phusion Human Specimen DNA Polymerase
Reaction Speed
Fast
Product Line
Phusion
Product Type
Direct PCR Kit
Hot Start
Built-In Hot Start
No. of Reactions
200 Reactions
Overhang
Blunt
Reaction Format
Separate Components
Sample Type
Human
Starting Material
DNA

Contents & Storage

Contains:
• 80 μL Phusion Human Specimen DNA Polymerase
• 2 x 1.25 mL 2X Phusion Human Specimen PCR Buffer (includes dNTPs and MgCl2)
• 40 μL Universal Control Primer Mix (25μM each)
• 2 x 5 mL Dilution Buffer
• 3 x 100 μL DNARelease Additive

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text