The FluoZin™-1 and FluoZin™-2 indicators are designed to detect Zn2+ concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists. Based on the N-(2-methoxyphenyl)iminodiacetate chelator, these indicators are designed for detection of Zn2+ in the 0-100 μM range with minimal interfering Ca2+ sensitivity. FluoZin™ indicators are offered in cell-permeant and cell-impermeant forms.
Fluorescent Zinc Indicators Specifications: • Label (Ex/Em): FluoZin™-2 (∼495/525 nm) • Kd (Zn2+) (in buffer): ∼2 μM • Lyophilized product may be dissolved in DMSO for use • Product is typically loaded into cells by adding the dissolved indicator to medium containing cells
Find Fluorescent Indicators for Zinc and Other Metal Ions We offer a number of fluorescent indicators to determine polyvalent cation concentrations inside cells, to follow metal ion transport through ion channels, to make measurements in environmental samples, and more. Review Fluorescent Indicators for Zn2+ and Other Metal Ions-Section 19.7 in the Molecular Probes™ Handbook for more information on these products.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)
Cell Viability and Proliferation
Product Type
Zinc Indicator
Product Line
FluoZin™
Shipping Condition
Room Temperature
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.