Invitrogen™

FISH Tag™ DNA Multicolor Kit, Alexa Fluor™ dye combination

Catalog number: F32951
Have Questions?
Invitrogen™

FISH Tag™ DNA Multicolor Kit, Alexa Fluor™ dye combination

Catalog number: F32951
Have Questions?
Catalog Number
F32951
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F329511 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The FISH Tag™ DNA Multicolor Kit provides a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. The FISH Tag™ DNA Multicolor Kit contains fluorescent dyes in four colors, making it ideal for simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and for in situ analysis of mitochondrial DNA localization and content. We also offer FISH Tag™ Kits in single colors.

FISH Tag™ Kit Specifications:
• Dye (Ex/Em): Alexa Fluor™ 488 (492/520 nm), Alexa Fluor™ 555 (555/565 nm), Alexa Fluor™ 594 (590/615 nm), Alexa Fluor™ 647 (650/670 nm)
• Comprehensive kit includes 16 reagents
• Comes with detailed protocol to help ensure your success


Complete Workflow Solution for FISH
FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. PureLink™ nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade™ Gold antifade reagent is added to preserve the fluorescent signal during imaging.

More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescence
Format
Kit
Label or Dye
Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647
Product Line
Alexa Fluor™, FISH Tag™
Product Type
DNA In Situ Hybridization Assay Kit
Quantity
1 kit
Target
DNA
Target Specificity
Not Target-Specific
Shipping Condition
Dry Ice
Unit Size
1 kit

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text