Invitrogen™

FluoroMyelin™ Red Fluorescent Myelin Stain - Solution in Water

Catalog number: F34652
Have Questions?
Invitrogen™

FluoroMyelin™ Red Fluorescent Myelin Stain - Solution in Water

Catalog number: F34652
Have Questions?
Catalog Number
F34652
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F346521 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The FluoroMyelin™ Red fluorescent myelin stain enables quick and selective labeling of myelin in brain cryosections in a single 20-minute labeling step plus washes. This stain can be used in conjunction with antibodies and other dyes, and with standard histochemical methods for cryosection material.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Red
Detection Method
Fluorescent
Product Type
Red Fluorescent Myelin Stain
Label or Dye
FluoroMyelin™ Red
Shipping Condition
Room Temperature
For Use With (Equipment)
Fluorescence Microscope
Sub Cellular Localization
Myelin
Excitation/Emission
558/654 nm
Product Line
FluoroMyelin™

Contents & Storage

• Provided as 300X concentration in water.
• When stored at 4°C, stable for at least 6 months after receipt.
• Protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text