Invitrogen™

FM™ 4-64FX, fixable analog of FM™ 4-64 membrane stain

Catalog number: F34653
Have Questions?
Invitrogen™

FM™ 4-64FX, fixable analog of FM™ 4-64 membrane stain

Catalog number: F34653
Have Questions?
Catalog Number
F34653
Unit Size
10 x 100 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F3465310 x 100 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The lipophilic probe, FM™ 4-64FX exhibits low fluorescence in water but fluoresces intensely upon binding the outer leaf of the plasma membrane providing discrete plasma membrane staining. The binding is rapid and reversible and 'FX' denotes that this analog is fixable with aldehyde-based fixatives. It is also an excellent reagent for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Infrared
For Use With (Equipment)
Fluorescence Microscope
Detection Method
Fluorescent
Product Line
FM™
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Lipophilic Probe
SubCellular Localization
Plasma Membrane

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text