Invitrogen™

FM™ 1-43FX, fixable analog of FM™ 1-43 membrane stain

Catalog number: F35355
Have Questions?
Invitrogen™

FM™ 1-43FX, fixable analog of FM™ 1-43 membrane stain

Catalog number: F35355
Have Questions?
Catalog Number
F35355
Unit Size
10 x 100 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F3535510 x 100 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
FM 1-43FX membrane probe is an FM 1-43 membrane probe analog that has been modified to contain an aliphatic amine. That modification makes the probe fixable with aldehyde-based fixatives (formaldehyde, glutaraldehyde, etc.). FM 1-43 membrane probe is widely used both for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling. Unmodified FM 1-43 membrane probe is available as 1 mg in a single vial (T-3163) and as 1 mg packaged into 10 vials, each containing 100 μg (T-35356).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Orange
For Use With (Equipment)
Fluorescence Microscope
Detection Method
Fluorescent
Product Line
FM™
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Membrane Stain
SubCellular Localization
Plasma Membrane

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text