The phospholipid, fluorescein DHPE is labeled on the head group with the green-fluorescent pH-sensitive fluorophore, fluorescein (excitation/emission maxima ∼496/519 nm).
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Chemical Name or Material
Phospholipids
Recommended Storage
Store in freezer (-5 to -30°C) and protect from light.
Quantity
5 mg
Physical Form
Solid
Product Line
Invitrogen
Figuras
Documentos e downloads
Certificados
Pesquisar por número de lote ou número de lote parcial