Thermo Scientific™

DyNAmo HS SYBR Green qPCR Kit

Catalog number: F410XL
Have Questions?
Thermo Scientific™

DyNAmo HS SYBR Green qPCR Kit

Catalog number: F410XL
Have Questions?
Catalog Number
F410XL
Unit Size
2,500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F410XL2,500 reactions
-
F410L500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of DyNAmo HS SYBR Green qPCR Kit and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific DyNAmo HS SYBR Green qPCR Kit contains a modified Thermus brockianus DNA polymerase and all the other reagents needed for qPCR. Only template DNA and PCR primers need to be added by the user. The modified DNA polymerase in this kit incorporates a non-specific DNA-binding domain that lends physical stability to the polymerase-DNA complex.

ROX passive reference dye is also supplied with the kit in a separate tube for instruments that require ROX normalization.

Highlights

• Strong specificity: DyNAmo HS SYBR Green master mix minimizes the formation of unwanted primer-dimers and extension of non-specifically bound primers during reaction setup
• Highly sensitive detection: Optimized master mix allows reproducible detection of low-copy number templates
• Broad dynamic range: DyNAmo HS SYBR Green qPCR Kit detects as few as one copy of the target gene and achieves a dynamic range of up to eight orders of magnitude
• Convenient to use: 2X master mix, one tube only. ROX passive reference dye provided in a separate vial

Related Products
DyNAmo Flash Probe qPCR Kit
DyNAmo Flash SYBR Green qPCR Kit
DyNAmo HS SYBR Green qPCR Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Tbr DNA polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
Kit Type
DyNAmo HS SYBR Green qPCR Kit
Product Line
DyNAmo
Product Type
Real Time PCR SYBR Master Mix
Sufficient For
2500 Reactions of 20 μL Each, 1000 Reactions of 50 μL Each
For Use With (Application)
Gene Expression
Concentration
2X (DyNAmo HS SYBR Green qPCR Mix), 50X (ROX Dye)
No. of Reactions
2500 Reactions
Sample Type
DNA, dsDNA
Volume
25 x 1 mL

Contents & Storage

Contains:
• 25 x1 mL DyNAmo HS SYBR Green qPCR Mix (2X)
• 1 x 1.25 mL 50 x ROX dye, 25μM

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text