Thermo Scientific™

DyNAmo Flash SYBR Green qPCR Kit

Catalog number: F415L
Have Questions?
Thermo Scientific™

DyNAmo Flash SYBR Green qPCR Kit

Catalog number: F415L
Have Questions?
Catalog Number
F415L
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F415L500 reactions
-
F415XL2,500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of DyNAmo Flash SYBR Green qPCR Kit and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific DyNAmo Flash SYBR Green qPCR Kit is developed for fast real-time qPCR. It provides qPCR results faster than most SYBR Green kits without compromising the qPCR performance. A DNA-binding domain attached to the polymerase in this kit improves the physical stability of the polymerase-DNA complex, thus enhancing the processivity and robustness of the amplification. The high amplification efficiency of DyNAmo Flash SYBR Green qPCR Kit gives reliable quantification and early Cq values. Due to the high signal intensity, DyNAmo Flash SYBR Green qPCR provides an excellent signal-to-noise ratio.

Highlights

• Extremely fast protocols: Combined annealing and extension step of only 15 s
• Specific and sensitive detection of a wide range of template concentrations
• Included dUTP allows the use of UNG for prevention of carry-over contamination
• Convenient 2× master mix and optimized protocol

Applications

• Fast qPCR
• qPCR using SYBR Green dye
• RT-qPCR using SYBR Green dye

Related Products
DyNAmo Flash Probe qPCR Kit
DyNAmo Flash SYBR Green qPCR Kit
DyNAmo HS SYBR Green qPCR Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Tbr DNA polymerase
Reaction Speed
Fast
For Use With (Equipment)
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
Kit Type
DyNAmo Flash SYBR Green qPCR Kit
Product Line
DyNAmo Flash
Product Type
Real Time PCR SYBR Master Mix
Sufficient For
500 Reactions of 20 μL, 200 Reactions of 50 μL
For Use With (Application)
Gene Expression
Concentration
2X (DyNAmo Flash SYBR Green qPCR Mix), 50X (ROX Dye)
No. of Reactions
500 reactions
Sample Type
DNA, dsDNA
Volume
5 x 1 mL

Contents & Storage

Contains:
• 5 x 1 mL DyNAmo Flash SYBR Green qPCR Mix (2X)
• 1 x 250 μL 50X ROX dye, 25μM

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text