Thermo Scientific™

DyNAmo ColorFlash SYBR Green qPCR Kit

Catalog number: F416XL
Have Questions?
Thermo Scientific™

DyNAmo ColorFlash SYBR Green qPCR Kit

Catalog number: F416XL
Have Questions?
Catalog Number
F416XL
Unit Size
2,500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
F416L
also known as F-416L
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F416XL2,500 reactions
-
F416L
also known as F-416L
500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of DyNAmo ColorFlash SYBR Green qPCR Kit and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific DyNAmo ColorFlash SYBR Green qPCR Kit offers equal performance to the DyNAmo Flash SYBR Green qPCR Kit. In addition, it incorporates an innovative multi-color system that ensures correct pipetting. The 2X master mix contains a blue dye, and a separate sample buffer contains a yellow dye. The qPCR reaction mix containing both components is green. Using this patent-pending multicolor system, pipetting of both the master mix and the sample can be easily monitored. This significantly decreases the risk for pipetting errors during reaction setup, especially when using white reaction vessels. The dyes do not affect the specificity or sensitivity of qPCR assays.

Highlights

• Colored reagents minimize the risk of pipetting errors during reaction set up
• Especially beneficial when using white reaction vessels
• Extremely fast protocols due to combined annealing and extension step of only 15 s
• Specific and sensitive detection of a wide range of template concentrations
• dUTP included in the 2x master mix allows the use of UNG for prevention of carry-over contamination

Applications

• Fast qPCR
• qPCR using SYBR Green dye
• RT-qPCR using SYBR Green dye

Related Products
DyNamo ColorFlash Probe qPCR Kit
DyNAmo ColorFlash SYBR Green qPCR Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Tbr DNA polymerase
Reaction Speed
Fast
For Use With (Equipment)
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
Kit Type
DyNAmo ColorFlash SYBR Green qPCR Kit
Product Line
DyNAmo Flash
Product Type
Real Time PCR SYBR Master Mix
Sufficient For
2500 Reactions of 20 μL, 1000 reactions of 50 μL
For Use With (Application)
Gene Expression
Concentration
2X (DyNAmo ColorFlash SYBR Green qPCR), 50X (ROX Dye), 40X (Sample Buffer Solution with Yellow Dye)
No. of Reactions
2500 Reactions
Sample Type
DNA, dsDNA
Volume
25 x 1 mL

Contents & Storage

Contains:
• 25 x 1 mL DyNAmo ColorFlash SYBR Green qPCR Mix (2X)
• 1 x 1.25 mL 50X ROX dye, 25μM
• 5 x 1 mL 40X sample buffer solution with yellow dye

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text