Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Varioskan LUX comes equipped with a range of measurement technologies including absorbance and fluorescence intensity with optional luminescence, AlphaScreen and TRF. The instrument selects the measurement wavelength using monochromators or optical filters, depending on which is optimal for each measurement technology. A wide range of optional filters extends the application area remarkably, including luciferase assays, Bioluminescence Resonance Energy Transfer (BRET) assays, Homogeneous Time Resolved Fluorescence (HTRF) assays and many other.
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 515 nm |
Excitation/Emission | 515/30 nm |
For Use With (Application) | BRET2 acceptor using GFP2 |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 460 nm |
Excitation/Emission | 460/80 nm |
For Use With (Application) | NanoBRET donor |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 480 nm |
Excitation/Emission | 480/20 nm |
For Use With (Application) | BRET donor using wtRluc |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 492 nm |
Excitation/Emission | 492/10 nm |
For Use With (Application) | General Terbium measurement |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 510 nm |
Excitation/Emission | 510/60 nm |
For Use With (Application) | Chroma-glo luciferase CBG68 or CBG99 assay |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 520 nm |
Excitation/Emission | 520/10 nm |
For Use With (Application) | LanthaScreen measurement of Tb acceptor; HTRF donor measurement of TbGreen |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 525 nm |
Excitation/Emission | 525/30 nm |
For Use With (Application) | Pierce green Renilla luciferase assay |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 530 nm |
Excitation/Emission | 530/30 nm |
For Use With (Application) | BRET acceptor using eYFP |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 571 nm |
Excitation/Emission | 571/77 nm |
For Use With (Application) | AlphaScreen measurement |
Unit Size | Each |
Product Type | Emission Filter |
For Use With (Equipment) | Varioskan LUX Microplate Reader |
Product Line | Varioskan™ |
Wavelength | 610 nm |
Excitation/Emission | 610/60 nm |
For Use With (Application) | Chroma-glo luciferase CBR assay |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Product Type | Wavelength | For Use With (Application) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
F51530 | Each | Emission Filter | 515 nm | BRET2 acceptor using GFP2 | 1,160.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F46080 | Each | Emission Filter | 460 nm | NanoBRET donor | 878.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F48020 | Each | Emission Filter | 480 nm | BRET donor using wtRluc | 1,160.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F49210 | Each | Emission Filter | 492 nm | General Terbium measurement | 986.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F51060 | Each | Emission Filter | 510 nm | Chroma-glo luciferase CBG68 or CBG99 assay | 1,160.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F52010 | Each | Emission Filter | 520 nm | LanthaScreen measurement of Tb acceptor; HTRF donor measurement of TbGreen | 986.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F52530 | Each | Emission Filter | 525 nm | Pierce green Renilla luciferase assay | 1,138.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F53030 | Each | Emission Filter | 530 nm | BRET acceptor using eYFP | 1,138.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F57177 | Each | Emission Filter | 571 nm | AlphaScreen measurement | 792.65 Online price 980.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
F61060 | Each | Emission Filter | 610 nm | Chroma-glo luciferase CBR assay | 1,160.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|