Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Larger pack sizes of Phusion Hot Start II DNA Polymerase with dNTPs now available.
Thermo Scientific™

Phusion™ Hot Start II DNA Polymerase (2 U/μL)

Catalog number: F549L
Have Questions?
Thermo Scientific™

Phusion™ Hot Start II DNA Polymerase (2 U/μL)

Catalog number: F549L
Have Questions?
Catalog Number
F549L
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F549L500 units
-
F549S100 units
-
F549XL2000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Using Phusion Hot Start II High-Fidelity DNA Polymerase, amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature.

The Affibody ligand also inhibits the 3´→5´ exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.

Highlights
• Reaction set up at room temperature
• No non-specific amplification and primer degradation during reaction set up
• Zero-time reactivation due to unique hot start technology
• High fidelity (52X Taq)
• Fast PCR due to short extension times (15-30 s/kb)
• Robust reactions, minimal optimization needed
• Increased product yields with minimal enzyme amounts
• Available as a Green buffer format permitting direct loading on gels (F-537S or F-537L)

Applications
• High-fidelity PCR
• High throughput
• Amplification of difficult (GC-rich) templates
• Template generation for sequencing
• Multiplex PCR
• Long-range PCR
• Cloning
• Mutagenesis
• Microarray

Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
Polymerase
Phusion Hot Start II DNA Polymerase
Reaction Speed
Fast
Product Type
Hot Start DNA Polymerase
Shipping Condition
Dry Ice
For Use With (Application)
Hot-start PCR, High-fidelity PCR
Concentration
2 U/μL
Fidelity (vs. Taq)
52X
Hot Start
Built-In Hot Start
No. of Reactions
500 Reactions
Overhang
Blunt
Reaction Format
Standalone
Size (Final Product)
20 kb or less

Contents & Storage

• Phusion Hot Start II High-Fidelity DNA Polymerase (2 U/μL)
• 5X Phusion HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text