Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phusion High-Fidelity PCR Kit

Catalog number: F553S
Have Questions?
Thermo Scientific™

Phusion High-Fidelity PCR Kit

Catalog number: F553S
Have Questions?
Catalog Number
F553S
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F553S50 reactions
-
F553L200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Highlights
• High fidelity (52X Taq)
• Fast PCR due to short extension times (15-30 s/kb)
• Robust performance, minimal optimization needed
• High yields of PCR products with minimal enzyme amounts
• Available as a Green buffer format for direct loading of PCR products on gels (F-534S or F-534L)

Applications
• High-fidelity PCR
• Cloning
• Template generation for sequencing
• Amplification of difficult (GC-rich) templates
• Long-range PCR (up to 20 kb)
• Mutagenesis
• High throughput PCR
• Microarray
Thermo Scientific Phusion High-Fidelity PCR Kit contains all the necessary reagents for PCR including control lambda DNA template and primers for 1.3 kb and 10 kb amplicons. The DNA template amount is sufficient for performing 20 control reactions in 50 μL volume or 50 control reactions in 20 μL volume.

Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
Polymerase
Phusion High-Fidelity DNA Polymerase
Reaction Speed
Fast
Product Type
High-Fidelity PCR Kit
Shipping Condition
Dry Ice
For Use With (Application)
Standard PCR, High-fidelity PCR
Fidelity (vs. Taq)
52X
Hot Start
No
No. of Reactions
50 Reactions
Overhang
Blunt
Reaction Format
Separate Components
Size (Final Product)
20 kb or less

Contents & Storage

• Phusion DNA Polymerase (2 U/μL)
• 5X Phusion GC & HF Buffer
• dNTP mix (10 mM each)
• Control template (λ DNA) and DNA size standard
• 1.3 kb & 10 kb primers (4 μM each)
• 50 mM MgCl2 solution and DMSO

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text