Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phusion U Hot Start DNA Polymerase

Catalog number: F555L
Have Questions?
Thermo Scientific™

Phusion U Hot Start DNA Polymerase

Catalog number: F555L
Have Questions?
Catalog Number
F555L
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F555L500 units
-
F555S100 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion, Phusion U overcomes an important limitation of proofreading enzymes - it is able to incorporate dUTP and read through uracil present in DNA templates.

In addition to uracil usage possibility, Phusion U features all the superior properties of other Phusion DNA Polymerases - great accuracy, speed, ability to amplify long amplicons up to 20 kb and a high specificity with Affibody based hot start. These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.

Phusion U Hot Start DNA Polymerase is also available in the Master Mix and Green formats. Phusion U Hot Start PCR Master Mix (F-533S and F-533L) is convenient 2X mix designed to minimize the number of pipetting steps. Only template and primers need to be added by the user. Phusion U Green format (F-556S and F-556L) is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Highlights
Accuracy—high fidelity DNA polymerase (25x Taq)
Uracil-tolerance—engineered to incorporate dUTP and amplify uracil containing templates
Specificity—hot start for reduced nonspecific amplification and primer degradation
Speed—short extension times (15-30 s/kb)

Applications
• Amplification of bisulphite-converted DNA
• Amplification of damaged or aged DNA
• Carry-over contamination control
• Uracil-excision based (USER) cloning methods

Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
Polymerase
Phusion U Hot Start DNA Polymerase
Reaction Speed
Fast
Product Type
Hot Start DNA Polymerase
Shipping Condition
Dry Ice
For Use With (Application)
Hot-start PCR, High-fidelity PCR
Concentration
2 U/μL
Fidelity (vs. Taq)
25X
Hot Start
Built-In Hot Start
No. of Reactions
500 Reactions
Overhang
Blunt
Reaction Format
Standalone
Size (Final Product)
20 kb or less

Contents & Storage

• Phusion U Hot Start DNA Polymerase, 2 U/μL
• 5X Phusion HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text