Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phusion U Multiplex PCR Master Mix

Catalog number: F562L
Have Questions?
Thermo Scientific™

Phusion U Multiplex PCR Master Mix

Catalog number: F562L
Have Questions?
Catalog Number
F562L
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F562L500 reactions
-
F562S100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Phusion U Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube. Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.

The Phusion U Multiplex PCR Master Mix contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Phusion U Green Multiplex PCR Master Mix (F-564S or F-564L) further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.

Highlights
• Specific and efficient multiplex PCR of over 20 targets up to 2.5 kb
• Excellent results with templates of suboptimal purity due to inhibitor tolerance
• Fast cycling and direct loading of PCR products on gels with Green format (F-564S and F-564L)

Applications
• Genotyping
• Pathogen detection
• Food testing
• Analysis of genetically modified organisms
• Amplification of microsatellites

The master mix is based on Phusion U Hot Start DNA Polymerase, a high performance enzyme developed using fusion technology. Similarly to other enzymes from Phusion family, Phusion U DNA Polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, Phusion U Multiplex PCR Master Mix can even be used with unpurified samples such as blood or serum.

Specificity of multiplex PCR is increased by Affibody-mediated hot start mechanism. Reversibly bound Affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.

Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
Medium
Polymerase
Phusion U Hot Start DNA Polymerase
Reaction Speed
Fast
Product Type
Multiplex PCR Master Mix
Shipping Condition
Dry Ice
For Use With (Application)
Hot-start PCR, Genotyping
Concentration
2X
Fidelity (vs. Taq)
25X
Hot Start
Built-In Hot Start
No. of Reactions
500 Reactions
Overhang
Blunt
Reaction Format
SuperMix or Master Mix
Size (Final Product)
2.5 kb or less

Contents & Storage

• Phusion U Hot Start DNA Polymerase
• 2X Reaction buffer
• 3 mM MgCl2
• 400 μM of each dATP, dTTP, dCTP and dGTP
• Nuclease-free water

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text