The FluoReporter™ FITC Protein Labeling Kit provides a convenient and flexible means for attaching fluorescein (FITC, Ex/Em ∼494/518 nm) to your monoclonal or polyclonal antibody (or any protein >30 kDa). The kit supplies the reagents needed for 5 to 10 labeling and separation reactions.
Important Features of the FluoReporter™ FITC Protein Labeling Kit:
• Labeled proteins typically ready to use in under 3 hours with very little hands-on time • Optimized for 0.2–2.0 mg of protein with molecular weight >30 kDa • Labeled proteins are purified using convenient spin filters • Requires the antibody or protein be purified prior to labeling
We’ll Make a Custom Labeled Protein for You If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom labeled protein for you. Our custom services are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
Labeling Kit
Quantity
1 kit
Detection Method
Fluorescence
Labeling Scale
0.2 to 2 mg
Shipping Condition
Room Temperature
Product Line
FluoReporter™
Label or Dye
FITC (Fluorescein)
Labeling Target
Antibodies, Proteins
Contents & Storage
Store in refrigerator 2°C to 8°C and protect from light.