Access Key Options for Ultra-Low Temperature Freezers
Access Key Options for Ultra-Low Temperature Freezers
Actual product may vary
Thermo Scientific™
Access Key Options for Ultra-Low Temperature Freezers
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FRAC34567Revco™ UxFField installed
ACE34567TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE SeriesAccess Key Pack (EU) Standalone Cards
ACU34567TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE SeriesAccess Key Pack (U.S.) Standalone Cards
FFAC34567Thermo Scientific TSU SeriesField installed
RAC34567TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE SeriesFactory installed. No stand-alone orders.
FAC34567LETSX Series, Revco RLE, HERAfreeze HLE, Forma 89000, TLE SeriesFactory installed. No stand-alone orders.
ELTSXUTSX UniversalFactory-Installed. No stand-alone orders
FELTSXUTSX UniversalField-installed
Catalog number FRAC34567
Price (USD)
-
Request A Quote
For Use With (Equipment):
Revco™ UxF
Type:
Field installed
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
  • Card access comes with 5 key cards
  • Field installed - purchased separately
  • Factory installed in the plant with purchase of unit
Specifications
For Use With (Equipment)Revco™ UxF
IncludesAccess Key Option; Card access control w/five key cards
TypeField installed
Unit SizeEach
Share catalog number, name or link