Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging
Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging
Invitrogen™
Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging
Biotinylated glutathione ethyl ester (BioGEE) is a cell-permeant, biotinylated glutathione analog for the detection of glutathiolation. Under conditions of oxidativeRead more
Have Questions?
Catalog NumberQuantity
G3600010 x 100 μg
Catalog number G36000
Price (USD)
305.00
Each
-
Add to cart
Quantity:
10 x 100 μg
Price (USD)
305.00
Each
Add to cart
Ask our AI about this Product
Biotinylated glutathione ethyl ester (BioGEE) is a cell-permeant, biotinylated glutathione analog for the detection of glutathiolation. Under conditions of oxidative stress, cells may transiently incorporate glutathione into proteins. Stressed cells incubated in BioGEE will also incorporate this biotinylated glutathione derivative into proteins, facilitating the identification of oxidation-sensitive proteins. Once these cells are fixed and permeabilized, you can detect glutathiolation levels with a fluorescent streptavidin conjugate using either flow cytometry or fluorescence microscopy. Proteins glutathiolated with BioGEE can also be extracted and analyzed by mass spectrometry or by Western blotting methods in conjunction with fluorophore- or enzyme-labeled streptavidin conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 x 100 μg
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link