Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Compatible Ladders | E-Gel™ Ultra Low Range DNA Ladder, Century™-Plus RNA Markers |
Gel Percentage | 4% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Separation Range | 20 to 800 bp |
Stain Type | SYBR™ Gold |
Wells | 11-well |
Mode of Separation | Molecular Weight |
Quantity | 10 Gels/Pk. |
Sample Type | Double-Stranded DNA (dsDNA), Single-Stranded RNA |
Shipping Condition | Room Temperature |
System Type | E-Gel™ |
Well Design | Single Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Gold |
Wells | 11-well |
Mode of Separation | Molecular Weight |
Quantity | 10 Gels/Pk. |
Sample Buffer Type | Preparative without dyes, Non-preparative |
Sample Type | Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA |
Shipping Condition | Room Temperature |
Well Design | Single Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Gold |
Wells | 11-well |
Mode of Separation | Molecular Weight |
Quantity | 20 Gels/Pk. |
Sample Buffer Type | Preparative without dyes, Non-preparative |
Sample Type | Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA |
Shipping Condition | Room Temperature |
Well Design | Single Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Century™-Plus RNA Markers |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Gold |
Wells | 11-well |
Mode of Separation | Molecular Weight |
Quantity | 10 Gels/Pk. |
Sample Buffer Type | Preparative without dyes, Non-preparative |
Sample Type | Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA |
Shipping Condition | Room Temperature |
Well Design | Single Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Century™-Plus RNA Markers |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Gold |
Wells | 11-well |
Mode of Separation | Molecular Weight |
Quantity | 20 Gels/Pk. |
Sample Buffer Type | Preparative without dyes, Non-preparative |
Sample Type | Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA |
Shipping Condition | Room Temperature |
Well Design | Single Comb |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Gel Percentage | Separation Range | Quantity | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
G401004 | Each | 4% | 20 to 800 bp | 10 Gels/Pk. | 249.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G401001 | Each | 1% | 100 to 5000 bp | 10 Gels/Pk. | 207.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G402021 | Each | 1% | 100 to 5000 bp | 20 Gels/Pk. | 343.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G401002 | Each | 2% | 50 to 2000 bp | 10 Gels/Pk. | 212.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G402022 | Each | 2% | 50 to 2000 bp | 20 Gels/Pk. | 346.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features of E-Gel EX agarose gels include:
Ultra-sensitive detection of DNA
E-Gel EX gels pre-stained with SYBR Gold DNA stain were developed for ultimate sensitivity and demonstrate up to 5-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.
Fast and convenient analysis
E-Gel EX gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 10–20 minutes. Each E-Gel agarose gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel straining steps. Just load your samples and run.
Live monitoring of DNA migration
The E-Gel EX agarose gels are pre-stained with SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, safe real-time monitoring of DNA or RNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.
Quick check of RNA samples
Besides DNA analysis, E-Gel EX gels can also be used for fast analysis of RNA samples to check their integrity before proceeding with downstream applications.
Easy access to the gel
E-Gel cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.
Running and imaging devices
E-Gel EX gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 11-well E-Gel EX agarose gels are available in 1%, 2%, and 4% gel percentages.