Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity | Separation Range | Gel Percentage |
---|---|---|---|
G402022 | 20 Gels/Pk. | 50 to 2000 bp | 2% |
G401001 | 10 Gels/Pk. | 100 to 5000 bp | 1% |
G402021 | 20 Gels/Pk. | 100 to 5000 bp | 1% |
G401002 | 10 Gels/Pk. | 50 to 2000 bp | 2% |
G401004 | 10 Gels/Pk. | 20 to 800 bp | 4% |
Features of E-Gel EX agarose gels include:
Ultra-sensitive detection of DNA
E-Gel EX gels pre-stained with SYBR Gold DNA stain were developed for ultimate sensitivity and demonstrate up to 5-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.
Fast and convenient analysis
E-Gel EX gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 10–20 minutes. Each E-Gel agarose gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel straining steps. Just load your samples and run.
Live monitoring of DNA migration
The E-Gel EX agarose gels are pre-stained with SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, safe real-time monitoring of DNA or RNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.
Quick check of RNA samples
Besides DNA analysis, E-Gel EX gels can also be used for fast analysis of RNA samples to check their integrity before proceeding with downstream applications.
Easy access to the gel
E-Gel cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.
Running and imaging devices
E-Gel EX gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 11-well E-Gel EX agarose gels are available in 1%, 2%, and 4% gel percentages.
Share catalog number, name or link