Invitrogen™

Safe Imager™ 2.0 Amber Filter Replacement

Catalog number: G6601
Have Questions?
Invitrogen™

Safe Imager™ 2.0 Amber Filter Replacement

Catalog number: G6601
Have Questions?
Catalog Number
G6601
Unit Size
1 unit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
G66011 unit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Safe Imager™ Blue-Light Transilluminator is designed for viewing stained gels on the laboratory bench top, or within gel documentation systems. Light from the LED source inside the instrument passes through a blue filter producing light with a narrow emission peak centered at approximately 470 nm, effective for the excitation of SYBR™ DNA-binding dyes such as SYBR™ Safe DNA gel stain, as well as many of our protein gel stains such as SYPRO™ Ruby, SYPRO™ Orange, and Pro-Q™ Diamond stains. Sensitivity obtained using this instrument is comparable to that obtained with a standard UV transilluminator. Unlike UV transilluminators, the Safe Imager™ transilluminator does not produce UV light and does not require UV protective equipment during use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Safe Imager™
Type
Filter Replacement
Quantity
1 unit
Shipping Condition
Room Temperature

Contents & Storage

Each instrument includes an amber filter unit and viewing glasses.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text