Informational:Discover our new Invitrogen E-Gel Agarose Gels with SYBR Safe DNA Stain—a safe alternative to ethidium bromide. Learn more »
Invitrogen™

E-Gel™ SizeSelect™ II Agarose Gels, 2%

Catalog number: G661012
Have Questions?
Invitrogen™

E-Gel™ SizeSelect™ II Agarose Gels, 2%

Catalog number: G661012
Have Questions?
Catalog Number
G661012
Unit Size
10 gels
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
G66101210 gels
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Electrophoresis using E-Gel SizeSelect II Agarose Gels is a convenient method for accurate size selection of DNA libraries for next-generation sequencing (NGS) applications. With E-Gel SizeSelect II gels, you can separate and recover DNA for library construction in three easy steps: load, run, and retrieve.

E-Gel SizeSelect II agarose gels enable you to:
Save time—no need to prepare a gel and gel-purify after electrophoresis
Simplify DNA recovery—retrieve purified DNA directly from the recovery well with a pipette
Improve result consistency—achieve accurate and consistent size selection

Fast and accurate DNA size selection in three easy steps
DNA fragment library size selection with E-Gel SizeSelect II precast gels is a simple 3-step procedure that eliminates conventional gel purification. To extract the DNA band of interest, load the sample into the loading wells, run the gel until the band reaches the recovery wells, and retrieve the sample. The resulting fragment library demonstrates a sharp fragment cut-off and is ready for use in an NGS workflow without any need for re-purification. Multiple bands from the one loading well can be collected during the same gel run.

DNA fragment libraries extracted from E-Gel SizeSelect II gels are compatible with the Ion Torrent sequencing workflow.

Real-time control of the sample
SYBR Gold II DNA Gel Stain is incorporated into the gel itself, allowing you to visually monitor DNA band migration progress on the blue-light transilluminator of the E-Gel Power Snap Electrophoresis Device. The gel has reference marks for better control of the sample prior to the sample recovery step. If the target DNA passes the recovery well, the Reverse gel protocol allows for re-capture of the DNA band.

Nucleic acid markers
Learn more about our E-Gel DNA ladders ›

Enables better results
Even a short exposure of your DNA sample to UV light during visualization may lead to DNA damage. Using E-Gel SizeSelect II gels together with the blue-light transilluminator minimizes the risk of UV damage compared to a conventional gel purification workflow. The direct access to your sample from the retrieval well allows you to avoid excising gel band slices and the risk of losing your DNA during a gel purification step.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Ladders
E-Gel™ Sizing DNA Ladder
Gel Percentage
2%
Gel Type
E-Gel
Product Type
Agarose Gel
Stain Type
SYBR™ Gold
Wells
7-well
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text