E-Gel™ 96 Agarose Gels
Discover our new Invitrogen E-Gel Agarose Gels with SYBR Safe DNA Stain—a safe alternative to ethidium bromide. Learn more »
E-Gel™ 96 Agarose Gels
Invitrogen™

E-Gel™ 96 Agarose Gels

Invitrogen E-Gel 96 high-throughput precast agarose gels with or without SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySeparation RangeStain TypeGel Percentage
G7008028 Gels100 to 2000 bpEthidium Bromide2%
G7008018 Gels400 to 10,000 bpEthidium Bromide1%
G720801Promo Image8 Gels400 to 10,000 bpSYBR™ Safe1%
G7208414 x 8 Gels400 to 10,000 bpSYBR™ Safe1%
G720802Promo Image8 Gels100 to 2000 bpSYBR™ Safe2%
G7208424 x 8 Gels100 to 2000 bpSYBR™ Safe2%
Catalog number G700802
Price (USD)
373.00
Special offer
Online exclusive
Ends: 21-Jul-2025
419.00
Save 46.00 (11%)
Each
Add to cart
Quantity:
8 Gels
Separation Range:
100 to 2000 bp
Stain Type:
Ethidium Bromide
Gel Percentage:
2%
Price (USD)
373.00
Special offer
Online exclusive
Ends: 21-Jul-2025
419.00
Save 46.00 (11%)
Each
Add to cart
Invitrogen E-Gel 96 high-throughput precast agarose gels with or without SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.

Invitrogen E-Gel 96 high-throughput precast agarose gels with or without SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.

Each gel contains 96 sample lanes and 8 marker lanes, ideal for screening multiple PCR products, plasmid preparations, and restriction digests. The E-Gel 96 loading format is compatible with multi-channel pipettors and the most commonly used 8-, 12-, and 96-pin liquid handling robots. With a 12-minute run time, up to 20,000 samples can be resolved in a single day.

The unique 96-well staggered-well format provides a 1.6-cm run length with the following separation ranges for DNA fragments:

  • 1% agarose gels: DNA fragments between 400 bp and 10 kb
  • 2% agarose gels: DNA fragments between 100 bp and 2 kb

Each cassette is labeled with an unique EAN39 bar code that is recognized by most commercially available robotic bar code readers.

Features of E-Gel 96 gels include:

  • High throughput analysis—contain 96 sample lanes and 8 marker lanes
  • Automation compatible—suitable for use with multi-channel pipettors and commonly used 8-, 12-, and 96-pin liquid handling robots
  • Scalable—resolve up to 384 samples in one run by combining up to four E-Base Mother and E-Base Daughter devices
  • Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
  • High performance—detect 3 ng of sample DNA with a separation time as short as 12–17 minutes

Safer option

Choose SYBR Safe DNA stained gels to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.

Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is safer for the user, safer for the environment, and can save institutional overhead costs for disposal.

Fast and convenient analysis

E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng of sample DNA per band and a separation time 2–3 fold faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a buffer-less ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.

Running and imaging devices

High-throughput E-Gel agarose gels require the E-Base Electrophoresis Device for gel running.

Available in a variety of gel formats

E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with or without SYBR Safe are available in 1% and 2% gel percentages.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeAgarose Gel
Quantity8 Gels
Shipping ConditionRoom Temperature
Stain TypeEthidium Bromide
Gel Percentage2%
Gel TypeE-Gel
Separation Range100 to 2000 bp
Wells96-well
Unit SizeEach
Contents & Storage
Each box contains 8 gels and a manual
  • Store at room temperature.

    Frequently asked questions (FAQs)

    My E-Gel agarose gel has speckles when viewed in the imager. What should I do?

    Here are some suggestions:

    - Try cleaning the cassettes with alcohol and Kimwipes wipers.
    - Try cleaning the camera lens.
    - Try to adjust the exposure time and brightness options of the documentation system you are using.

    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

    My sample is leaking from the wells when running my E-Gel agarose gels. What happened?

    Please ensure that you have not overloaded the well and that the wells were not damaged during comb removal.

    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

    I accidentally stored my E-Gel agarose gels at 4 degrees C instead of room temperature. Can I still use them?

    While we recommend storage at room temperature, these gels will still be usable. Bring the gels to room temperature prior to the run for optimal conditions.

    Find additional tips, troubleshooting help, and resources within ourNucleic Acid Gel Electrophoresis and Blotting Support Center.

    Are there robotic scripts for running E-Gel 48/96 or E-PAGE 48/96 gels?

    Robotic scripts for running E-Gel 96 Agarose Gels and E-PAGE 96 Gels on Beckman Coulter's Biomek FX workstation can be found on our website at: www.thermofisher.com/egels (click the Labware Definitions link in the left navigation pane).

    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

    What loading buffer should I use for my E-Gel agarose gels?

    Loading buffer is optional. Samples can be loaded directly into the wells if no buffer is used, or you can dilute them with deionized water or TE buffer. If you want to use a loading buffer, please see the recipes below:

    E-Gel agarose gels (including EX)
    10 mM Tris-HCl, pH 7.5
    1 mM EDTA
    0.005% bromophenol blue
    0.005% xylene cyanol FF
    E-Gel CloneWell II and E-Gel SizeSelect II agarose gels
    10 mM Tris-HCl, pH 7.5
    1 mM EDTA

    Alternatively, you can use 10X BlueJuice Gel Loading Buffer or TrackIt Loading Buffer. Dilute this buffer 50- to 200-fold to obtain optimal results with E-Gel agarose gels.

    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

    Documents & Downloads

    Certificates

    Safety Data Sheets

    Share catalog number, name or link