E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 1%
Build your own complete bundle and get personalized pricing! Learn more now.
E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 1%
Invitrogen™
E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 1%
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, EX 1%, provides everything you need to quickly start using precastRead more
Have Questions?
Catalog NumberQuantity
G8341ST1 kit
Catalog number G8341ST
Price (USD)
4,085.00
Each
In stock
Add to cart
Quantity:
1 kit
Price (USD)
4,085.00
Each
Add to cart
Ask our AI about this Product
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, EX 1%, provides everything you need to quickly start using precast E-Gel EX 1% agarose gels on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
  • 10 E-Gel EX agarose gels, 1% (Cat. No. G401001)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel 1 Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
  • Gel opening knife (Cat. No. EI9010)
Learn more about the E-Gel Power Snap Electrophoresis System ›
Learn more about precast E-Gel EX agarose gel technology ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Shipping ConditionRoom Temperature
Voltage240 VAC
Gel SizeMini
Max. Voltage48 VDC
TypeElectrophoresis System Starter Kit
Unit SizeEach
Contents & Storage
• 1 electrophoresis device
• 1 camera device
• 10 E-Gel EX 1% gels
• 1 vial E-Gel 1 Kb Plus Express DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter
• 1 knife
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection