Thermo Scientific™

Gemini™ Chemical Identification Analyzers

Catalog number: GEMINI
Have Questions?
Thermo Scientific™

Gemini™ Chemical Identification Analyzers

Catalog number: GEMINI
Have Questions?
Identify a broad range of unknown chemicals and explosives in the field using FTIR and Raman spectroscopy in a single instrument, including low concentration analysis. The rugged, compact Thermo Scientific™ Gemini™ analyzer meets the demanding requirements of elite military and public safety forces, helping operators execute their mission quickly, safely, and accurately.
 
Catalog Number
GEMINI
Unit Size
Each
Includes
5 years of support: reachback support, software updates and library updates
Price (USD)
Full specifications
Product TypeHandheld Raman/FTIR Spectrometer
Certifications/ComplianceCertified to MIL-STD-810G and IP67; full test report available on request
Collection OpticsFTIR: ATR diamond crystal, single bounce
Raman: NA= 0.33
Data Export FormatsSPC file (for use in standard spectroscopic software), text file, or JPEG report
DescriptionGemini Analyzer
Dimensions (L x W x H)10.1 x 5.7 x 2.4 in. (25.6 x 14.6 x 6.1 cm)
Includes5 years of support: reachback support, software updates and library updates
IP RatingIP67
LanguagesEnglish
Power SupplyInternal 3.7 V lithium polymer (QTY 1) battery pack
Internal CR123A (QTY 3) batteries
DC wall adapter, 12 V DC, 1.25 A
Spectral RangeFTIR: 4,000 cm-1 to 650 cm-1
Raman: 250 cm-1 to 2875 cm-1
Temperature Range (Metric)-20°C to +50°C
Weight (English)4.2 lb.
Weight (Metric)1.9 kg
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeIncludesPrice (USD)
GEMINIFull specifications
Each5 years of support: reachback support, software updates and library updatesRequest A Quote
Product TypeHandheld Raman/FTIR Spectrometer
Certifications/ComplianceCertified to MIL-STD-810G and IP67; full test report available on request
Collection OpticsFTIR: ATR diamond crystal, single bounce
Raman: NA= 0.33
Data Export FormatsSPC file (for use in standard spectroscopic software), text file, or JPEG report
DescriptionGemini Analyzer
Dimensions (L x W x H)10.1 x 5.7 x 2.4 in. (25.6 x 14.6 x 6.1 cm)
Includes5 years of support: reachback support, software updates and library updates
IP RatingIP67
LanguagesEnglish
Power SupplyInternal 3.7 V lithium polymer (QTY 1) battery pack
Internal CR123A (QTY 3) batteries
DC wall adapter, 12 V DC, 1.25 A
Spectral RangeFTIR: 4,000 cm-1 to 650 cm-1
Raman: 250 cm-1 to 2875 cm-1
Temperature Range (Metric)-20°C to +50°C
Weight (English)4.2 lb.
Weight (Metric)1.9 kg
Unit SizeEach
Showing 1 of 1

The Gemini Analyzer is the first and only integrated Raman and FTIR handheld instrument in the world, providing complementary and confirmatory testing in a single, field-portable device.


Better, faster decision making

  • Lightweight and compact, ideal for chemical battalions, explosive ordnance disposal (EOD) technicians and other first responders burdened with heavy equipment.
  • Virtual mixture analysis and interactive problem solving on scene with HazMasterG3.
  • Includes 24/7 reachback support with access to Ph.D. spectroscopists for spectral analysis and operational inquiries.


Increased confidence in results

  • Leverages both Raman and FTIR technology to address a broader range of samples than either technique alone.  
  • Identifies unknown solids and liquids, from explosives and chemical warfare agents to industrial chemicals and precursors using a comprehensive onboard library. 
  • Builds on the field-proven performance of Thermo Scientific™ FirstDefender™ and Thermo Scientific™ TruDefender™ Analyzers.


Safer operation

  • Includes a motorized anvil which adjusts sample pressure based on user settings.
  • Features Raman and FTIR Scan Delay to allow users to leave the hazard zone before initiating a scan.


Easier to use, enhanced efficiency

  • Guides the operator through technology selection and sampling with Scan Assist.
  • Customizable Profiles allow the operator to set scan parameters based on the sample, scenario and their situational awareness
  • Minimizes user error and eliminates subjective user interpretation.
  • Requires minimal training for proficiency.

 

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text