Search Thermo Fisher Scientific
Type | Greenhouse Gas Analyzer |
Detection Mode | Flame Ionization, Electron Capture, Thermal Conductivity |
Column Capacity | 6 |
Description | 3-channel gas chromatograph with configured valve oven |
Includes | TRACE 1310 GC with one FID detector, one ECD detector, one TCD detecor, Auxiliary valve oven with (2) 10-port valves, 4 columns, and complete documentation package. System is fully assembled and factory tested. |
Function | Automated analysis of a gas sample for green house gases, CH4, CO2, and N2O. |
Detector Type | FID, ECD, TCD |
Unit Size | Each |
Type | Greenhouse Gas Analyzer |
Detection Mode | Flame Ionization and Electron Capture |
Column Capacity | 6 |
Description | Dual-channel gas chromatograph with configured valve oven |
Includes | TRACE 1310 GC with one FID detector, one ECD detector, Auxiliary valve oven with (2) 10-port valves, 4 columns, a methanizer and complete documentation package. System is fully assembled and factory tested. |
Function | Automated analysis of a gas sample for greenhouse gases, CH4, CO2, and N2O. |
Detector Type | FID and ECD |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Includes | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
GHA000010021 | Each | 3-channel gas chromatograph with configured valve oven | TRACE 1310 GC with one FID detector, one ECD detector, one TCD detecor, Auxiliary valve oven with (2) 10-port valves, 4 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GHA000010011 | Each | Dual-channel gas chromatograph with configured valve oven | TRACE 1310 GC with one FID detector, one ECD detector, Auxiliary valve oven with (2) 10-port valves, 4 columns, a methanizer and complete documentation package. System is fully assembled and factory tested. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|