Search Thermo Fisher Scientific
Obtain presumptive identification of Gram negative organisms in just five hours, or obtain species level identification with overnight incubation using the Thermo Scientific™ Sensititre™ Gram Negative GNID ID Plate. If extended incubation is required for identification, the same plate can be re-incubated, eliminating unnecessary waste.
Description | Sensititre Gram Negative Identification Plate |
Product Type | Identification Test |
Target Organism Class | Gram-negative Microorganisms |
Test Type | Presumptive Identification |
Isolates Per Plate | 3 |
No. per Pack | 10 |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
GNID | Each | Sensititre Gram Negative Identification Plate | 10/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.