Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
0.5 McFarland Standard | H2O (Part No. T3339) |
Antimicrobial Agents | Various |
AST Type | MIC |
Broth Type | MHB (Part No. T3462) |
Description | Sensititre Gram Negative Urine Plate |
Antibiotic Code | Various |
Product Type | MIC Plate |
Target Organism Class | Gram-negative Microorganisms |
Test Type | Minimum Inhibitory Concentration (MIC) |
Incubation Conditions | 35°C O2 |
Incubation Time | 18 hr |
Isolates Per Plate | 3 |
No. per Pack | 10 |
Quantity | 10/Pk. |
Reading Options | Autoread or manual |
Transfer Volume | For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL |
Volume Per Well | 50 μL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
GNUR2F | Each | Sensititre Gram Negative Urine Plate | 10/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|