Search Thermo Fisher Scientific
The Thermo Scientific™ GS Omni Light Element Analyzer provides robust, accurate elemental analysis for multiple process streams, delivering real-time information for the optimization of slurry beneficiation processes such as froth flotation. Utilizing industry-leading Prompt Gamma Neutron Activation Analysis (PGNAA) technology it measures both heavy and light elements, crucially covering the Z<20 elements - calcium and lighter - that are less well-suited to X-Ray Fluorescence (XRF) analysis. Together, Thermo Scientific PGNAA and XRF analyzers allow customers to measure the broadest possible range of elements.
Since the PGNAA signal is unaffected by particle size, ore matrix or inter-element interference the GS Omni requires no recalibration as the ore changes, enabling robust measurement with minimal cost and ongoing effort. Easily configured for between two and eight process streamsit provides data that you can confidently rely on. Use the GS Omni to control contaminant elements, improve product quality, increase recovery, and lower production costs.
Description | The analyzer is available in configurations of 1 to 8 streams. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
GSOMNI | Each | The analyzer is available in configurations of 1 to 8 streams. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The GS Omni offers near metallurgical quality analysis for up to eight process streams, depending on concentration, elements of interest, required cycle analysis time, plant layout and process control considerations.
Key features include: