Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RRHRFSEEEL KPQPIMKKAR KIQVPEEQKD EKYWSRRYKN NEAAKRSRDA RRLKENQISV RAAFLEKENA LLRQEVVAVR QELSHYRAVL SRYQAQHGAL
D-site-binding protein (DBP) is a member of the PAR bZIP (proline and acidic amino acid-rich basic leucine zipper) transcription factor family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Albumin D box-binding protein; Albumin D-element-binding protein; D site of albumin promoter (albumin D-box) binding protein; D site-binding protein; Tax-responsive enhancer element-binding protein 302; TaxREB302
Gene Aliases: DABP; DBP
UniProt ID: (Human) Q10586
Entrez Gene ID: (Human) 1628
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support