The HCS DNA Damage kit simultaneously and quantitatively measures two important cell-health parameters: DNA damage and cytotoxicity. DNA damage isRead more
Have Questions?
Catalog Number
Quantity
H10292
1 kit
Catalog number H10292
Price (EUR)
556,00
Each
-
Add to cart
Quantity:
1 kit
Price (EUR)
556,00
Each
Add to cart
The HCS DNA Damage kit simultaneously and quantitatively measures two important cell-health parameters: DNA damage and cytotoxicity. DNA damage is detected using an antibody against phosphorylated H2AX (Ser139) which is induced in response to double-strand break (DSB) formation. To detect cytotoxicity, the Image-iT® DEAD™ Green is an impermeant dye in healthy cells that becomes permeant when the plasma membrane of cells is compromised. Both indicators are amenable to fixation and permeabilization, thus allowing for multiplexing with other biomarkers of interest. The blue-fluorescent nuclear segmentation tool Hoechst 33342 is also included and stains DNA in both live and dead cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAlexa Fluor™ 555, Hoechst 33258
Format96-well plate
Quantity1 kit
Shipping ConditionDry Ice
ColorOrange, Blue, Green
For Use With (Equipment)High Content Analysis Instrument
Product LineMolecular Probes
Product TypeDNA Damage kit
Unit SizeEach
Contents & Storage
The kit contains sufficient reagents for performing 2 plates in a 96-well plate format.
Store at ≤-20°C, desiccate and protect from light.