The HCS Mitochondrial Health Kit simultaneously and quantitatively measures two important cell-health parameters: mitotoxicity and cytotoxicity. The MitoHealth stain detects changes in the mitochondrial membrane potential, whereupon the reagent accumulates in active mitochondria. The kit includes the Image-iT® DEAD™ Green to detect cytotoxicity, an impermeant dye to healthy cells that becomes permeant when the plasma membrane of cells is compromised. Both indicators are amenable to fixation and permeabilization, thus allowing for multiplexing with other biomarkers of interest. The blue-fluorescent nuclear segmentation tool Hoechst 33342 is also included and stains DNA in both live and dead cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Color
Orange, Blue, Green
Detection Method
Fluorescent
For Use With (Equipment)
Fluorescence Microscope, High Content Analysis Instrument
Product Type
Mitochondrial Health Kit
Dye Type
Image-iT™ DEAD Green viability stain, MitoHealth stain, Hoechst 33342
Emission
UV, Visible
Format
96-well plate
Product Line
Molecular Probes™
Shipping Condition
Room Temperature
Contents & Storage
The kit contains sufficient reagents for performing 2 plates in a 96-well plate format.
Store at ≤-20°C, desiccate and protect from light.