Thermo Scientific™

Hypoxia Green Reagent for Flow Cytometry

Catalog number: H20035
Have Questions?
Thermo Scientific™

Hypoxia Green Reagent for Flow Cytometry

Catalog number: H20035
Have Questions?
Catalog Number
H20035
Unit Size
2 x 50 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
H200352 x 50 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Hypoxia Green Reagent for Flow Cytometry is an antibody-independent reagent used to detect low levels of oxygen levels in live cells. The membrane-permeant probe releases rhodamine as oxygen levels decrease, resulting in a fluorogenic response. Hypoxia is a characteristic of many diseases, including cardiovascular disease and tumor-mediated immunosuppression and is critical to tumor survival and growth.

• Sensitive, fixed, end-point assay
• Detects decreases in oxygen in live cells
• Fluorogenic, non-antibody based probe

Specifications

Detection Method
Fluorescent
For Use With (Equipment)
Flow Cytometer
Product Type
Reagent
Emission
488
Format
Vial(s)
Quantity
2 x 50 assays
Shipping Condition
Room Temperature

Contents & Storage

2 vials of 50 tests each
  • Store at -5°C to -30°C.
  • Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text