Thermo Scientific Chemicals

2,2-Difluorocyclopropanecarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H25752.06
5 g, Each
Thermo Scientific Chemicals

2,2-Difluorocyclopropanecarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H25752.06
5 g, Each
Quantity
Catalog number: H25752.06
also known as H25752-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107873-03-0
IUPAC Name
2,2-difluorocyclopropane-1-carboxylic acid
Molecular Formula
C4H4F2O2
InChI Key
KMLMOVWSQPHQME-UHFFFAOYNA-N
SMILES
OC(=O)C1CC1(F)F
Appearance (Color)
White to very pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Assay (GC)
≥94.0%
Melting Point (clear melt)
59.0-65.0?C

Description

2,2-Difluorocyclopropanecarboxylic acid derivative (i.e. 2,2-difluorocyclopropanecarbonyl chloride) is used in Friedel-Crafts reactions with various arenes to prepare aryl 2,2-difluorocyclopropyl ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorocyclopropanecarboxylic acid derivative (i.e. 2,2-difluorocyclopropanecarbonyl chloride) is used in Friedel-Crafts reactions with various arenes to prepare aryl 2,2-difluorocyclopropyl ketones.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text