Thermo Scientific Chemicals

2-Methoxy-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: H25772.03
1 g, Each
Thermo Scientific Chemicals

2-Methoxy-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: H25772.03
1 g, Each
Quantity
Catalog number: H25772.03
also known as H25772-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63071-03-4
IUPAC Name
2-methoxy-6-methylpyridine
Molecular Formula
C7H9NO
InChI Key
WIMNZMOEBDPZTB-UHFFFAOYSA-N
SMILES
COC1=CC=CC(C)=N1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4975-1.5015 @ 20?C

Description

2-Methoxy-6-methylpyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxy-6-methylpyridine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text