2-Bromo-6-fluoroaniline, 98+%, Thermo Scientific Chemicals
2-Bromo-6-fluoroaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-6-fluoroaniline, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H25774.06
also known as H25774-06
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.30 
Save 12.65 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.30 
Save 12.65 (15%)
Add to cart
2-Bromo-6-fluoroaniline, 98+%, Thermo Scientific Chemicals
Catalog numberH25774.06
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.30 
Save 12.65 (15%)
-
Add to cart
Chemical Identifiers
CAS65896-11-9
IUPAC Name2-bromo-6-fluoroaniline
Molecular FormulaC6H5BrFN
InChI KeyALZFPYUPNVLVQM-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
Refractive Index1.5795-1.5835 @ 20?C
FormLiquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only