7-Benzyloxyindole, 98%, Thermo Scientific Chemicals
7-Benzyloxyindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Benzyloxyindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H25776.03
also known as H25776-03
Price (USD)/ Each
36.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
36.80
Add to cart
7-Benzyloxyindole, 98%, Thermo Scientific Chemicals
Catalog numberH25776.03
Price (USD)/ Each
36.80
-
Add to cart
Chemical Identifiers
CAS20289-27-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to re brown
FormCrystalline powder
Assay (GC)>97.5%
Comment4-Benzyloxyindole; <0.1%
CommentToluene: <0.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only