DL-Cysteine hydrochloride, 90+%, Thermo Scientific Chemicals
DL-Cysteine hydrochloride, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Cysteine hydrochloride, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number H25779.06
also known as H25779-06
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
Add to cart
DL-Cysteine hydrochloride, 90+%, Thermo Scientific Chemicals
Catalog numberH25779.06
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10318-18-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥90.0 to ≤110.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only