Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H25791.06
also known as H25791-06
Price (USD)/ Each
201.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
201.00
Add to cart
Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate, 98%, Thermo Scientific Chemicals
Catalog numberH25791.06
Price (USD)/ Each
201.00
-
Add to cart
Chemical Identifiers
CAS33545-44-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)>97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only