Thermo Scientific Chemicals

(1-Butyl)triphenylphosphonium chloride, Thermo Scientific Chemicals

Catalog number: H25796.22
100 g, Each
Thermo Scientific Chemicals

(1-Butyl)triphenylphosphonium chloride, Thermo Scientific Chemicals

Catalog number: H25796.22
100 g, Each
Quantity
Catalog number: H25796.22
also known as H25796-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13371-17-0
IUPAC Name
butyltriphenylphosphanium chloride
Molecular Formula
C22H24ClP
InChI Key
MFIUDWFSVDFDDY-UHFFFAOYSA-M
SMILES
[Cl-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Powder
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Water Content (Karl Fischer Titration)
≤1.0%
Melting Point
224-230?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text