Thermo Scientific Chemicals

Acetoxyacetyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H25797.06
5 g, Each
Thermo Scientific Chemicals

Acetoxyacetyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H25797.06
5 g, Each
Quantity
Catalog number: H25797.06
also known as H25797-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13831-31-7
IUPAC Name
2-chloro-2-oxoethyl acetate
Molecular Formula
C4H5ClO3
InChI Key
HZDNNJABYXNPPV-UHFFFAOYSA-N
SMILES
CC(=O)OCC(Cl)=O
Refractive Index
1.4245-1.4295 @ 20°C (non-U.S. sourced material)
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Comment
Material Sourced in the U.S. and in other countries

Description

Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams and glycolylhydroxamic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams and glycolylhydroxamic acids.

Solubility
Reacts with water violently.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text