2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H25822.06
also known as H25822-06
Price (USD)/ Each
277.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
277.00
Add to cart
2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
Catalog numberH25822.06
Price (USD)/ Each
277.00
-
Add to cart
Chemical Identifiers
CAS175205-82-0
IUPAC Name2-bromo-3-(trifluoromethyl)pyridine
Molecular FormulaC6H3BrF3N
InChI KeyOFGSIPQYQUVVPL-UHFFFAOYSA-N
SMILESFC(F)(F)C1=C(Br)N=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)37.0-43.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only