Thermo Scientific Chemicals

2,2-Difluoro-N-methoxy-N-methylacetamide, 95%, Thermo Scientific Chemicals

Catalog number: H25833.06
5 g, Each
Thermo Scientific Chemicals

2,2-Difluoro-N-methoxy-N-methylacetamide, 95%, Thermo Scientific Chemicals

Catalog number: H25833.06
5 g, Each
Quantity
Catalog number: H25833.06
also known as H25833-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142492-01-1
IUPAC Name
2,2-difluoro-N-methoxy-N-methylacetamide
Molecular Formula
C4H7F2NO2
InChI Key
CUPRFYMJGQMIIC-UHFFFAOYSA-N
SMILES
CON(C)C(=O)C(F)F
Assay (GC)
≥94.0%
Refractive Index
1.3990-1.4050 @ 20?C
Appearance (Color)
Clear colorless to yellow or orange to brown
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only