(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%, Thermo Scientific Chemicals
(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H25854.03
also known as H25854-03
Price (USD)/ Each
232.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
232.00
Add to cart
(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%, Thermo Scientific Chemicals
Catalog numberH25854.03
Price (USD)/ Each
232.00
-
Add to cart
Chemical Identifiers
CAS119580-41-5
IUPAC Name2-(trifluoromethyl)pyrrolidine
Molecular FormulaC5H8F3N
InChI KeyVINAMCOZNJHNIH-UHFFFAOYNA-N
SMILESFC(F)(F)C1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormFused solid or clear liquid as melt or clear liquid
Assay (GC)≥94.0%
Optical Rotation+9.5° to +11° (c=1 in methanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only