4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals
4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H25855.03
also known as H25855-03
Price (USD)/ Each
296.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
296.00
Add to cart
4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals
Catalog numberH25855.03
Price (USD)/ Each
296.00
-
Add to cart
Chemical Identifiers
CAS17573-78-3
IUPAC Name4,5,6-trifluoropyrimidine
Molecular FormulaC4HF3N2
InChI KeyAQKOJKCYBNUFLU-UHFFFAOYSA-N
SMILESFC1=NC=NC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4055-1.4105 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only