Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals
Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number H25864.14
also known as H25864-14
Price (USD)/ Each
145.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
145.00
Add to cart
Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberH25864.14
Price (USD)/ Each
145.00
-
Add to cart
Chemical Identifiers
CAS545375-30-2
IUPAC Name2-aminoethyl benzoate hydrochloride
Molecular FormulaC9H12ClNO2
InChI KeyZXTMLOSVFWKYCL-UHFFFAOYSA-N
SMILESCl.NCCOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Assay (HPLC)≥98.0%
Melting Point (clear melt)138.5-144.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only