2-(Ethoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
2-(Ethoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Ethoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H25867.03
also known as H25867-03
Price (USD)/ Each
25.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
25.90
Add to cart
2-(Ethoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberH25867.03
Price (USD)/ Each
25.90
-
Add to cart
Chemical Identifiers
CAS380430-53-5
IUPAC Name[2-(ethoxycarbonyl)phenyl]boronic acid
Molecular FormulaC9H11BO4
InChI KeyQZKVVOXAEBCLPZ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to yellow
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only