Chlorobis(4-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals
Chlorobis(4-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorobis(4-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H25887.06
also known as H25887-06
Price (USD)/ Each
366.65
Online exclusive
407.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
366.65
Online exclusive
407.00 
Save 40.35 (10%)
Add to cart
Chlorobis(4-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals
Catalog numberH25887.06
Price (USD)/ Each
366.65
Online exclusive
407.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13685-30-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to plae cream or pale grey
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥95.0%
Proton NMRConforms to structure
Chlorobis(4-methoxyphenyl)phosphine is used an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(4-methoxyphenyl)phosphine is used an pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.
RUO – Research Use Only

General References:

  1. Masanobu Yatagai; Takamichi Yamagishi; Mitsuhiko Hida. Asymmetric Hydrogenation of Dehydrodipeptides with Rhodium(I)-Chiral Diphosphinites. Selective (S,S)- and (R,R)-Product Formation by Double Asymmetric Induction. Bulletin of the Chemical Society of Japan. 1984, 57 (3), 823-826.
  2. X. Ren; L. Wang and Y. Li. 1,1'-Bis[bis(4-methoxyphenyl)phosphanyl]ferrocene. Acta Cryst. 2012, 68, 922.