Thermo Scientific Chemicals

Chlorodi(p-tolyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: H25889.03
1 g, Each
Thermo Scientific Chemicals

Chlorodi(p-tolyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: H25889.03
1 g, Each
Quantity
Catalog number: H25889.03
also known as H25889-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1019-71-2
IUPAC Name
chlorobis(4-methylphenyl)phosphane
Molecular Formula
C14H14ClP
InChI Key
BJBXRRHIBSXGLF-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)P(Cl)C1=CC=C(C)C=C1
Proton NMR
Conforms to structure
Appearance (Color)
Clear, colorless to pale yellow
Form
Viscous liquid
Assay by NMR
≥94.0%
Phosphorus-31 NMR
Conforms to structure

Description

Chlorodi(p-tolyl)phosphine is used as a reactant for synthesis of palladium catalysts for C-C bond forming cross-coupling reactions, ligands for nickel(0)-catalyzed isomerization reactions, phosphinosulfonamide nickel complexes for oligomerization reactions. It is a reactant for phosphinylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodi(p-tolyl)phosphine is used as a reactant for synthesis of palladium catalysts for C-C bond forming cross-coupling reactions, ligands for nickel(0)-catalyzed isomerization reactions, phosphinosulfonamide nickel complexes for oligomerization reactions. It is a reactant for phosphinylation reactions.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text